Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIKE-L/S Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07675MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-675-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-675-MI Supplier MilliporeSigma Supplier No. 07675

Rabbit Polyclonal Antibody

Detect PIKE-L/S using this Anti-PIKE-L/S Antibody validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIKE-L/S
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols CENTG1; GGAP2; PIKE; KIAA0167; FLJ16430; AGAP-2; AGAP2
Host Species Rabbit
Immunogen 6His-tagged fusion protein corresponding to residues 268-384 of rat PIKE-L
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014770.2
Target Species Human, Rat
Content And Storage −20°C for up to 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.