Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pin1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07091MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-091-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-091-MI Supplier MilliporeSigma Supplier No. 07091
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Pin1 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Pin1 and has been validated in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pin1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Presentation: 100 μg of protein A purified rabbit IgG in 100μL of 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% Sodium Azide and 30% glycerol. Liquid at −20°C.
Gene Symbols PIN1; dod; UBL5; DOD
Host Species Rabbit
Immunogen Full-length His tagged human Pin-1
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006221
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.