Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pinch-1 Mouse anti-Human, Mouse, Clone: 9D2.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT162
Description
Anti-Pinch-1 Antibody, clone 9D2.1 is a highly specific mouse monoclonal antibody, that targets Pinch-1 and has been tested in western blotting and immunohistochemistry.
Specifications
Pinch-1 | |
Monoclonal | |
Unconjugated | |
NP_001180411 | |
Mouse | |
Purified by ion-exchange chromatography | |
Primary | |
2-8°C for one year | |
IgM κ |
Immunohistochemistry, Western Blot | |
9D2.1 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
LIMS1, PINCH, PINCH1 | |
KLH-conjugated linear peptide corresponding to human Pinch-1. | |
100 μg | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction