Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB100644
Description
PINK1 Polyclonal specifically detects PINK1 in Human samples. It is validated for Western Blot, Simple Western.Specifications
PINK1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
PINK1 | |
PINK1 antibody was developed using a synthetic peptide made residues 258-274 (YRKSKRGPKQLAPHPNI) of human PINK1. | |
Affinity Purified | |
RUO | |
Primary | |
Reacts with residues residues 258-274 (YRKSKRGPKQLAPHPNI) of human PINK1 and will only bind to isoform 1. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot | |
1 mg/mL | |
Western Blot 1:100-1:2000, Simple Western 1:50 | |
BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mitochondrial | |
Rabbit | |
62.7 kDa | |
0.1 mg | |
Alzheimers Research, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
65018 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction