Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PINK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals BC100494SS
Description
PINK1 Polyclonal specifically detects PINK1 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA, Immunoassay, KnockDown.Specifications
| PINK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Anti-PINK1, BRPK, EC 2.7.11.1, FLJ27236, Mitochondrial, PARK6, Parkinson disease (autosomal recessive) 6, PINK1, PINK1 mouse, PINK1 polyclonal, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protei | |
| Rabbit | |
| 62.7 kDa | |
| 0.025 mL | |
| Alzheimers Research, Cardiovascular Biology, DNA Repair, Lipid and Metabolism, Mitochondrial Markers, Mitophagy, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| 65018 | |
| Human, Mouse, Rat, Drosophila | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ELISA, Immunoassay, KnockDown | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation reported in scientific literature (PMID 22078885), Immunohistochemistry-Paraffin reported in scientific literature (PMID 25083992), Immunohistochemistry-Frozen reported in scientific literature (PMID 31908016), Immunoblotting reported in multiple pieces of scientific literature, Peptide ELISA 1:100 - 1:2000, Electron Microscopy, SDS-Page reported in scientific literature (PMID 27846363), Knockout Validated reported in scientific literature (PMID 31066324), KnockDown Validated | |
| Q9BXM7 | |
| PINK1 | |
| PINK1 antibody was developed using a synthetic peptide made to the human PINK1 protein sequence (between residues 175-250). [Swiss-Prot Q9BXM7] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human PINK1 Antibody will be reactive to isoform 2. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction