Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphatidylinositol-5-phosphate 4-kinase, type II, alpha, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017954 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-954 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-954 Supplier Abnova Corporation Supplier No. H00005305D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PIP4K2A protein.

Phosphatidylinositol-5,4-bisphosphate, the precursor to second messengers of the phosphoinositide signal transduction pathways, is thought to be involved in the regulation of secretion, cell proliferation, differentiation, and motility. The protein encoded by this gene is one of a family of enzymes capable of catalyzing the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. The amino acid sequence of this enzyme does not show homology to other kinases, but the recombinant protein does exhibit kinase activity. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family. [provided by RefSeq

Sequence: MATPGNLGSSVLASKTKTKKKHFVAQKVKLFRASDPLLSVLMWGVNHSINELSHVQIPVMLMPDDFKAYSKIKVDNHLFNKENMPSHFKFKEYCPMVFRNLRERFGIDDQDFQNSLTRSAPLPNDSQARSGARFHTSYDKRYIIKTITSEDVAEMHNILKKYHQYIVECHGITLLPQFLGMYRLNVDGVEIYVIVTRNVFSHRLSVYRKYDLKGSTVAREASDKEKAKELPTLKDNDFINEGQKIYIDDNNKKVFLEKLKKDVEFLAQLKLMDYSLLVGIHDVERAEQEEVECEENDGEEEGESDGTHPVGTPPDSPGNTLNSSPPLAPGEFDPNIDVYGIKCHENSPRKEVYFMAIIDILTHYDAKKKAAHAAKTVKHGAGAEISTVNPEQYSKRFLDFIGHILT

Specifications

Antigen phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PIP4K2A protein.
Formulation PBS with no preservative; pH 7.4
Gene PIP4K2A
Gene Accession No. NM_005028.3
Gene Alias FLJ13267/PI5P4KA/PIP5K2A/PIP5KII-alpha/PIP5KIIA/PIPK
Gene Symbols PIP4K2A
Host Species Rabbit
Immunogen PIP4K2A (NP_005019.2, 1 a.a. ∼ 406 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5305
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.