Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIR-A/B Rat anti-Mouse, Unlabeled, Clone: 6C1, BD

Catalog No. BDB550348 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550348 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550348 Supplier BD Biosciences Supplier No. 550348
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6C1 antibody reacts with common epitopes of PIR-A and PIR-B (Paired Immunoglobulin-liked Receptors) in all mouse strains tested (A/J, BALB/cJ, C3H, C57BL/6, DBA/1, DBA/2, NZB, and SJL). PIR-A and PIR-B are type-I transmembrane glycoproteins containing six Ig-like domains. There are multiple PIR-A proteins which are activating receptors by virtue of their intracellular association with the ITAM (Immunoreceptor Tyrosine-based Activation Motif)- containing Fc receptor γ chain (FcRγ) in mast cells and macrophages. FcRγ expression is required for PIR-A cell-surface expression on dendritic cells, B cells, and myeloid lineages. In contrast, PIR-B is an inhibitory receptor which contains various ITIMs (Immunoreceptor Tyrosine-based Inhibitory Motifs) in its cytoplasmic domain. These receptors are expressed on B cells, granulocytes, mast cells, dendritic cells, and monocytes/macrophages, but not on thymocytes, T lymphocytes, erythroid lineage cells, or NK cells. The level of cell-surface expression of these receptors increases as a function of B-cell activation and myeloid- and B-lineage differentiation. The 6C1 antibody immunoprecipitates molecules of 85- and 125kDa, which correspond to PIRA and PIR-B, respectively.

Host Species: Rat
Clone: 6C1
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant mouse PIR-A1 extracellular domain peptide and the mouse myeloid cell line, WEH13

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen PIR-A/B
Applications Flow Cytometry
Classification Monoclonal
Clone 6C1
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Recombinant mouse PIR-A1 extracellular domain peptide and the mouse myeloid cell line, WEH13
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.