Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIR121-1/Sra-1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-531-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-531-MI Supplier MilliporeSigma Supplier No. 07531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PIR121-1/Sra-1 Antibody is an antibody against PIR121-1/Sra-1 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIR121-1/Sra-1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols CYFIP2; KIAA1168; PIR121
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 872-886 (SQEFQRDKQPNAQPQ-C) of human PIR121-1/Sra-1, with a C-terminal cysteine added for conjugation purposes.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_014608
Target Species Human, Mouse
Content And Storage −70°C for up to 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.