Learn More
PITPNA, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PITPNA peptide using ARM Technology.
Supplier: Abnova Corporation H00005306K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of a family of lipid-binding proteins that transfer molecules of phosphatidylinositol or phosphatidylcholine between membrane surfaces. The protein is implicated in phospholipase C signaling and in the production of phosphatidylinositol 3,4,5-trisphosphate (PIP3) by phosphoinositide-3-kinase
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PITPNA | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PITPNA | |
PITPNA | |
A synthetic peptide of human PITPNA | |
RUO | |
5306 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
phosphatidylinositol transfer protein, alpha | |
PITPNA | |
MGC99649, PITPN, VIB1A | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.