Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

piwi-like 1 (Drosophila), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016970 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-970 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-970 Supplier Abnova Corporation Supplier No. H00009271B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PIWIL1 protein.

This gene encodes a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. The encoded protein may play a role as an intrinsic regulator of the self-renewal capacity of germline and hematopoietic stem cells. [provided by RefSeq

Sequence: MTGRARARARGRARGQETAQLVGSTASQQPGYIQPRPQPPPAEGELFGRGRQRGTAGGTAKSQGLQISAGFQELSLAERGGRRRDFHDLGVNTRQNLDHVKESKTGSSGIIVRLSTNHFRLTSRPQWALYQYHIDYNPLMEARRLRSALLFQHEDLIGKCHAFDGTILFLPKRLQQKVTEVFSKTRNGEDVRITITLTNELPPTSPTCLQFYNIIFRRLLKIMNLQQIGRNYYNPNDPIDIPSHRLVIWPGFTTSILQYENSIMLCTDVSHKVLRSETVLDFMFNFYHQTEEHKFQEQVSKELIGLVVLTKYNNKTYRVDDIDWDQNPKSTFKKADGSEVSFLEYYRKQYNQKITDLKQPVLVSQPKRRRGPGGTLPGPAMLIPELCYLTGLTDKMRNDFNVMKDLAVHTRLTPEQRQREVGRLIDYIHKNDNVQRELRDWGLSFDSNLLSFSGRILQTEKIHQGGKTFDYNPQFADWSKETRGAPLISVKPLDNWLLIYTRRNYEAANSLIQNLFKVTPAMGMQMRKAIMIEVDDRTEAYLRVLQQKVTADTQIVVCLLSSNRKDKYDAIKKYLCTDCPTPSQCVVARTLGKQQTVMAIATKIALQMNCKMGGELWRVDIPLKLVMIVGIDCYHDMTAGRRSIAGFVASINEGMTRWFSRCIFQDRGQELVDGLKVCLQAALRAWNSCNEYMPSRIIVYRDGVGDGQLKTLVNYEVPQFLDCLKSIGRGYNPRLTVIVVKKRVNTRFFAQSGGRLQNPLPGTVIDVEVTRPEWYDFFIVSQAVRSGSVSPTHYNVIYDNSGLKPDHIQRLTYKLCHIYYNWPGVIRVPAPCQYAHKLAFLVGQSIHREPNLSLSNRLYYL

Specifications

Antigen piwi-like 1 (Drosophila)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PIWIL1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PIWIL1
Gene Accession No. NM_004764
Gene Alias HIWI/MIWI/PIWI
Gene Symbols PIWIL1
Host Species Mouse
Immunogen PIWIL1 (NP_004755, 1 a.a. ∼ 861 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9271
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.