Learn More
Phospho-PKA alpha/beta (Thr197), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA128490
Description
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The protein encoded by this gene is a member of the Ser/Thr protein kinase family and is a catalytic subunit of cAMP-dependent protein kinase. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
PA1-28490 detects PKA alpha + beta from mouse, human samples.PA1-28490 has been successfully used in Western blot applications.The PA1-28490 immunogen is: Synthetic phosphopeptide derived from a region of human PKA that contains threonine 197.
Specifications
Phospho-PKA alpha + beta pThr197 | |
Polyclonal | |
Unconjugated | |
P00517, P05131, P17612, P22694, P05132, P68181, P05383, P36887, P27791, P68182 | |
PRKACA, PRKACB | |
Synthetic phosphopeptide derived from a region of human PKA that contains threonine 197. | |
50 μL | |
Primary | |
Bovine, Human, Mouse, Pig, Rat | |
Antibody | |
IgG |
Western Blot | |
0.35mg/mL | |
PRKACA | |
PRKC Pan, Protein kinase C alpha type, PKC-A, PKC-alpha, PRKCA, PKCA, PRKACA, PKCB, PRKCB1, Protein kinase C beta type, PKC-B, PKC-beta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5566, 5567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.