Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA[C] Mouse, Unlabeled, Clone: 5B, BD

Catalog No. BDB610981 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610981 150 μg
BDB610980 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610981 Supplier BD Biosciences Supplier No. 610981
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic (C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα, and RIIβ. These subunits define type I and II cAMP-dependent protein kinases. Following binding of cAMP, the regulatory subunits dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Type II PKA can be distinguished by autophosphorylation of the R-subunits, while type I PKA binds Mg/ATP with high affinity. The levels of expression of the different subunits vary according to cell and tissue type.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen PKA[C]
Applications Western Blot
Classification Monoclonal
Clone 5B
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human PKA[Cα] subunit aa. 18-347
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2b
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.