Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA Rabbit anti-Bovine, Human, Hamster, Mouse, Pig, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06903MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-903-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-903-MI Supplier MilliporeSigma Supplier No. 06903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-PKA Antibody, NT detects level of PKA and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol added to 30%
Gene Symbols PRKACA; MGC102831; PKACA; MGC48865
Host Species Rabbit
Immunogen peptide (KKGSEQESVKEFLAK-C) based upon amino acids 8-22 of the human 41kDa alpha isoform of the catalytic subunit of PKA
Purification Method Protein A chromatography
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002730
Target Species Bovine, Hamster, Human, Mouse, Pig, Rat
Content And Storage −20°C for 24 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.