Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC Mouse anti-Human, Mouse, Rat, Clone: M110, MilliporeSigma™

Catalog No. 05983MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-983-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-983-MI Supplier MilliporeSigma Supplier No. 05983
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PKC (α Antibody, β, γ), clone M110 detects level of PKC (α and has been published and validated for use in ELISA, immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC (α, β, γ)
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M110
Conjugate Unconjugated
Formulation 100μL of protein A purified mouse monoclonal IgG1 in 50% affinity storage buffer (PBS containing 1.0mg/mL BSA and 0.05% sodium azide) and 50% glycerol.
Gene Accession No. NP_002728, NP_002730
Gene Symbols PRKCA, PKC-A, AAG6, PKC-alpha, MGC129901, MGC129900, PRKACA, PKCA
Host Species Mouse
Immunogen Recombinant protein of human PKCγ amino acids 499-697.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.