Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCΕ Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 06991MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-991-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-991-MI Supplier MilliporeSigma Supplier No. 06991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect PKCΕ using this Anti-PKCΕ Antibody validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKCε
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide, and 30% glycerol.
Store at −20°C.
Gene Symbols PRKCE; MGC125656; PKCE; MGC125657; nPKC-epsilon
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to the C-terminal variable (V5) region, amino acids 726-737 (KGFSYFGEDLMP), of mouse Protein Kinase C Epsilon (PKCε). The immunizing sequence is identical in human, mouse, rat and rabbit.
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_005400
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.