Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-PKCzeta Clone: 4B12.3,

Catalog No. MABC946MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC946MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC946MI Supplier MilliporeSigma™ Supplier No. MABC946
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects PKCζ Clone: 4B12.3 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Protein kinase C zeta type (EC 2.7.11.13; UniProt Q05513; also known as nPKC-zeta, PKC-zeta) is encoded by the PRKCZ (also known as PKC2) gene (Gene ID 5590) in human. Protein kinase Cs (PKCs) are threonine/serine kinases that play important roles in cellular signal transductions. PKC isozymes are classified into three major classes based on their domain compositions and activator requirements. The conventional PKCs (cPKCs) include the alpha, beta I, beta II, and gamma isoforms. The novel PKCs (nPKCs) include the delta, epsilon, eta, and theta isoforms. Atypical PKCs (aPKCs) include the lambda (mouse ortholog to human PKC-lambda is called PKC-iota) and zeta isoforms. Both cPKCs and nPKCs bind diacylglycerol (DAG) via their C1 domain, while only cPKCs, but not nPKCs bind Ca+2 via their C2 domain. The aPKCs do not bind either DAG or calcium for activity. All three classes of PKCs contain a pseudosubstrate sequence in their N-terminal region that keeps the substrate-binding cavity occupied via intramolecular interaction when the enzymes are in their inactive conformation. Conformation change upon enzyme activation frees the substrate-binding site.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKCζ
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B12.3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q05513
Gene Symbols PRKCZ, PKC2
Host Species Mouse
Immunogen GST-tagged recombinant human PKCζ C-terminal fragment.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002735
Test Specificity Expected to react with all three spliced isoforms of human PKC-zeta.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.