Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHH2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526776
Description
Specifications
| PLEKHH2 | |
| Polyclonal | |
| PLEKHH2 | |
| PLEKHH1L, pleckstrin homology domain-containing family H member 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 130271 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q8IVE3 | |
| PLEKHH2 | |
| KLH conjugated synthetic peptide between 1049-1079 amino acids from the C-terminal region of human PKHH2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction