Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN2 Mouse anti-Human, Rat, Mouse, Monkey, Clone: 1D1, MilliporeSigma™

Catalog No. MABT496MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT496MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT496MI Supplier MilliporeSigma Supplier No. MABT496
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PKN2, clone 1D1 Antibody is an antibody against PKN2 for use in western blotting, ELISA, immunohistochemistry, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKN2
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1D1
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols PKN2, PRK2, PRKCL2
Host Species Mouse
Immunogen Purified recombinant fragment of human PKN2 expressed in E. Coli
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.