Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PBX/knotted 1 homeobox 1, Mouse, Clone: 4F6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant PKNOX1.
Supplier: Abnova Corporation H00005316M01
Description
Sequence: MATQTLSIDSYQDGQQMQVVTELKTEQDPNCSEPDAEGVSPPPVESQTPMDVDKQAIYRHPLFPLLALLFEKCEQSTQGSEGTTSASFDVDIENFVRKQSpecifications
PBX/knotted 1 homeobox 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004571 | |
PKNOX1 | |
PKNOX1 (NP_004562, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Transcription Regulation | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
4F6 | |
Mouse monoclonal antibody raised against a partial recombinant PKNOX1. | |
PKNOX1 | |
PREP1/pkonx1c | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
5316 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction