Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phosphatidylinositol 4-kinase type 2 alpha, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-519 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-519 Supplier Abnova Corporation Supplier No. H00055361D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PI4K2A protein.

Phosphatidylinositolpolyphosphates (PtdInsPs) are centrally involved in many biologic processes, ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4KII phosphorylates PtdIns at the D-4 position, an essential step in the biosynthesis of PtdInsPs (Barylko et al., 2001 [PubMed 11244087]).[supplied by OMIM

Sequence: MDETSPLVSPERAQPPDYTFPSGSGAHFPQVPGGAVRVAAAAGSGPSPPGSPGHDRERQPLLDRARGAAAQGQTQTVAAQAQALAAQAAAAAHAAQAHRERNEFPEDPEFEAVVRQAELAIERCIFPERIYQGSSGSYFVKDPQGRIIAVFKPKNEEPYGHLNPKWTKWLQKLCCPCCFGRDCLVLNQGYLSEAGASLVDQKLELNIVPRTKVVYLASETFNYSAIDRVKSRGKRLALEKVPKVGQRFNRIGLPPKVGSFQLFVEGYKDADYWLRRFEAEPLPENTNRQLLLQFERLVVLDYIIRNTDRGNDNWLIKYDCPMDSSSSRDTDWVVVKEPVIKVAAIDNGLAFPLKHPDSWRAYPFYWAWLPQAKVPFSQEIKDLILPKISDPNFVKDLEEDLYELFKKDPGFDRGQFHKQIAVMRGQILNLTQALKDNKSPLHLVQMPPVIVETARSHQRSSSESYTQSFQSRKPFFSWW

Specifications

Antigen phosphatidylinositol 4-kinase type 2 alpha
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PI4K2A protein.
Formulation PBS with no preservative; pH 7.4
Gene PI4K2A
Gene Accession No. NM_018425
Gene Alias DKFZp761G1923/PI4KII/PIK42A/RP11-548K23.6
Gene Symbols PI4K2A
Host Species Rabbit
Immunogen PI4K2A (NP_060895.1, 1 a.a. ∼ 479 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55361
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.