Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G1B, Mouse, Clone: 1B3, Abnova™

Catalog No. 89024810 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-810 Supplier Abnova Corporation Supplier No. H00005319M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PLA2G1B.

Phospholipase A2 (EC 3.1.1.4) catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets (Seilhamer et al., 1986 [PubMed 3028739]) .[supplied by OMIM

Sequence: SGISPRAVWQFRKMIKCVIPGSDPFLEYNNYGCYCGLGGSGTPVDELDKQKQRV

Specifications

Antigen PLA2G1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant PLA2G1B.
Formulation PBS with no preservative; pH 7.4
Gene PLA2G1B
Gene Accession No. BC005386
Gene Alias MGC119834/MGC119835/PLA2/PLA2A/PPLA2
Gene Symbols PLA2G1B
Host Species Mouse
Immunogen PLA2G1B (AAH05386, 17 a.a. ∼ 70 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5319
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.