Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plakophilin 2 Mouse anti-Human, Unlabeled, Clone: 28, BD

Catalog No. BDB610788 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610788 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610788 Supplier BD Biosciences Supplier No. 610788
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plakophilin 2 is a desmosomal plaque protein that belongs to the “arm” repeat family of proteins. Alternative splicing results in two isoforms: 2a (837 aa) and 2b (881 aa). The 2b isoform contains an insertion of 44 aa in the arm repeat unit 2. Other arm family members are β-catenin, plakophilin 1, plakoglobin, pp120, and armadillo of Drosophila gene product. They have at least 9 repeats of the arm motif, which consists of approximately 42 aa, and a short carboxyl terminus. Plakophilins are constitutively expressed in desmosome-containing cells. Plakophilin 1 has been found only in desmosomal-containing cells, such as stratified and complex epithelia. Plakophilin 2 has been isolated from both desmosomal as well as non-desmosomal forming cells, including simple and glandular epithelia, and from nonepithelial tissues such as the mycocardium and Purkinje fibers. In addition, Plakophilin 2 is found in the karyoplasm of interphase cells. Plakophilins 2a and 2b are part of desmosomal plaques, although they are located primarily in the karyoplasm of the interphase nucleus. The function of nuclear plakophilin 2 remains to be identified.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Plakophilin 2
Applications Western Blot
Classification Monoclonal
Clone 28
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Plakophilin 2a aa. 19-183
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.