Learn More
Urokinase, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA19084
Description
Human tPA produced in CHO cells is a single, glycosylated polypeptide chain containing 527 amino acids and having a molecular mass of 59008.71 Dalton. rHutPA is a serine protease enzyme that converts plasminogen to plasmin. The rHutPA is purified by proprietary chromatographic techniques.
Specifications
| Urokinase | |
| Polyclonal | |
| Unconjugated | |
| P00749 | |
| PLAU | |
| Synthetic peptide corresponding to the C terminus amino acids HFLPWIRSHTKEEN. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.5mg/mL | |
| PLAU | |
| PLAU, plasminogen activator, urokinase, HGNC:9052, ATF, UPA, URK, u-PA , U-plasminogen activator, antagonist of uPA, plasminogen activator, urinary, u | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5328 | |
| Store at -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.