Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasminogen/Angiostatin Mouse anti-Human, Clone: GMA-013, MilliporeSigma™

Catalog No. 05862MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-862-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-862-MI Supplier MilliporeSigma Supplier No. 05862
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Plasminogen/Angiostatin Antibody, clone GMA-013 is validated for use in ELISA, western blotting for the detection of Plasminogen/Angiostatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plasminogen/Angiostatin
Applications ELISA, Western Blot
Classification Monoclonal
Clone GMA-013
Conjugate Unconjugated
Gene Accession No. NM_000301
Gene Symbols PLG, DKFZp779M0222, plasminogen
Host Species Mouse
Immunogen Purified human plasminogen
Purification Method DEAE purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 24 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.