Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCγ-1 Mouse anti-Bovine, Human, Mouse, Rat, Rabbit, Clone: B-6-4, MilliporeSigma™

Catalog No. 05366MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-366-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-366-MI Supplier MilliporeSigma Supplier No. 05366
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PLCγ-1 Antibody, clone B-6-4 is an antibody against PLCγ-1 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCγ-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone B-6-4
Conjugate Unconjugated
Formulation Lyophilized powder
Gene Accession No. NM_002660
Gene Symbols PLCG1, PLC1, PLCgamma1, PLC148, PLC-II, PLC-148, 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PLC-gamma-1, inositoltrisphosphohydrolase
Host Species Mouse
Immunogen Purified soluble phospholipase C (PLC) g-1 from bovine brain
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rabbit, Rat
Content And Storage Lyophilized: 2 years at −20°C; Rehydrated: 6 months at −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.