Learn More
PLCγ-1 Mouse anti-Human, Mouse, Rat, Rabbit, Bovine, Clone: Clonal blend B-2-5, B-6-4, B-20-3, D-7-3, E-9-4., MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | PLCγ-1 |
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | Clonal blend B-2-5, B-6-4, B-20-3, D-7-3, E-9-4., B-6-4, B-20-3, D-7-3, E-9-4. |
| Conjugate | Unconjugated |
| Formulation | Liquid in 0.1MTris-glycine, pH 7.4, 0.15MNaCl and 0.05% sodium azide. An equal mix of IgG antibodies produced by SP2/0-Ag14 x Balb/c spleen cells (Suh, P.G., 1988) and propagated as ascites. Clones B-2-5, B-6-4, B-20-3, D-7-3, E-9-4. |
| Gene Accession No. | NM_002660.2 |
| Gene Symbols | PLCG1, PLC1, PLCgamma1, PLC148, PLC-II, PLC-148, 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PLC-gamma-1, inositoltrisphosphohydrolase |
| Host Species | Mouse |
| Immunogen | Five individual clones raised against purified soluble phospholipase C (PLC)γ-1 from bovine brain. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.