Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLC-γ2 Mouse, Alexa Fluor 647, Clone: K86-1161, BD

Catalog No. BDB560136 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560136 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560136 Supplier BD Biosciences Supplier No. 560136

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol biphosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLC-γ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with receptor tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: 1) PLC-γ1, which is ubiquitously expressed, and 2) PLCγ2, found primarily in the lymphoid system. PLC-γ is essential for growth factor-induced cell motility and mitogenesis. Overexpression of PLC-γ is evident in several forms of cancer, and it has been identified as a key mediator of PDGF-dependent cellular transformation. Thus regulation of PLC-γ activity by growth factors is involved in cell growth and transformation. Although the immunogen for generation of the K86-1161 monoclonal antibody was a phosphorylated peptide, peptide blocking studies demonstrated that the mAb recognizes PLC-γ2 regardless of phosphorylation status. This antibody was raised to a unique region of PLC-γ2 and is predicted not to crossreact with PLC-γ1.

Host Species: Mouse
Clone: K86-1161
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Phosphorylated Human PLC-γ2 Peptide

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLC-γ2
Applications Flow Cytometry
Classification Monoclonal
Clone K86-1161
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human PLC-γ2 Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.