Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLC-γ Mouse anti-Human, Alexa Fluor 647, Clone: 10/PLCGAMMA, BD

Catalog No. BDB558565 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558565 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558565 Supplier BD Biosciences Supplier No. 558565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Phospholipase C (PLC) isozymes hydrolyze phosphatidyl inositol biphosphate to inositol triphosphate and diacylglycerol. The former causes release of calcium from the endoplasmic reticulum, while the latter is an activator of Protein Kinase C. Within the PLC family, PLCγ is the only member that contains SH2 and SH3 domains. These domains enable it to interact with receptor tyrosine kinases and become enzymatically activated via phosphorylation. It exists as two isoforms: 1) PLCγ1, which is ubiquitously expressed, and 2) PLCγ2, found primarily in the lymphoid system. PLCγ is essential for growth factor-induced cell motility and mitogenesis. PLCγ1 null mice exhibit retarded embryonic growth and lethality in midgestation. Overexpression of PLCγ is evident in several forms of cancer, and it has been identified as a key mediator of PDGF-dependent cellular transformation. Thus regulation of PLCγ activity by growth factors is involved in cell growth and transformation. The 10/PLCgamma monoclonal antibody recognizes PLCγ1, regardless of phosphorylation status. It does not cross-react with PLCγ2.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLC-γ
Applications Flow Cytometry
Classification Monoclonal
Clone 10/PLCGAMMA
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Cow PLCγ1 N-terminal region Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.