Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phospholipase C, delta 4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008365 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-365 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-365 Supplier Abnova Corporation Supplier No. H00084812B01

Mouse polyclonal antibody raised against a full-length human PLCD4 protein.

Phosphatidylinositol-specific phospholipase C (PLC) plays an important role in receptor-mediated signal transduction by generating 2 second messenger molecules, inositol 1,4,5-triphosphate (IP3) and diacylglycerol, from phosphatidylinositol 4,5-bisphosphate (PIP2). PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 [PubMed 8381210]).[supplied by OMIM

Sequence: MASLLQDQLTTDQDLLLMQEGMPMRKVRSKSWKKLRYFRLQNDGMTVWHARQARGSAKPSFSISDVETIRNGHDSELLRSLAEELPLEQGFTIVFHGRRSNLDLMANSVEEAQIWMRGLQLLVDLVTSMDHQERLDQWLSDWFQRGDKNQDGKMSFQEVQRLLHLMNVEMDQEYAFSLFQAADTSQSGTLEGEEFVQFYKALTKRAEVQELFESFSADGQKLTLLEFLDFLQEEQKERDCTSELALELIDRYEPSDSGKLRHVLSMDGFLSYLCSKDGDIFNPACLPIYQDMTQPLNHYFICSSHNTYLVGDQLCGQSSVEGYIRALKRGCRCVEVDVWDGPSGEPVVYHGHTLTSRILFKDVVATVAQYAFQTSDYPVILSLETHCSWEQQQTMARHLTEILGEQLLSTTLDGVLPTQLPSPEELRRKILVKGKKLTLEEDLEYEEEEAEPELEESELALESQFETEPEPQEQNLQNKDKKKKSKPILCPALSSLVIYLKSVSFRSFTHSKEHYHFYEISSFSETKAKRLIKEAGNEFVQHNTWQLSRVYPSGLRTDSSNYNPQELWNAGCQMVAMNMQTAGLEMDICDGHFRQNGGCGYVLKPDFLRDIQSSFHPEKPISPFKAQTLLIQVISGQQLPKVDKTKEGSIVDPLVKVQIFGVRLDTARQETNYVENNGFNPYWGQTLCFRVLVPELAMLRFVVMDYDWKSRNDFIGQYTLPWTCMQQGYRHIHLLSKDGISLRPASIFVYICIQEGLEGDES

Specifications

Antigen phospholipase C, delta 4
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PLCD4 protein.
Formulation No additive
Gene PLCD4
Gene Accession No. NM_032726
Gene Alias MGC12837
Gene Symbols PLCD4
Host Species Mouse
Immunogen PLCD4 (NP_116115, 1 a.a. ∼ 762 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Endocrine/Metabolism
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84812
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.