Learn More
PLD1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PLD1 peptide using ARM Technology.
Supplier: Abnova Corporation H00005337K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995 [PubMed 8530346]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| PLD1 | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| PLD1 | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 1mg | |
| phospholipase D1, phosphatidylcholine-specific | |
| PLD1 | |
| PLD1 | |
| A synthetic peptide of human PLD1 | |
| RUO | |
| 5337 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.