Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLD4/Phospholipase D4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP213770
Description
PLD4/Phospholipase D4 Polyclonal specifically detects PLD4/Phospholipase D4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PLD4/Phospholipase D4 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| C14orf175, Choline phosphatase 4, chromosome 14 open reading frame 175, EC 3.1.4.4, Phosphatidylcholine-hydrolyzing phospholipase D4, phospholipase D family, member 4, phospholipase D4, PLD 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PLD4 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: AQDLEKTFQTYWVLGVPKAVLPKTWPQNFSSHFNRFQPFHGLFDGVPTTAYFSASPPALCPQGRTRDLEALLAVMGSAQEFIYASVMEYFP | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 122618 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction