Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plectin Mouse anti-Rat, Human, Mouse, Clone: 10F6, MilliporeSigma™

Catalog No. MAB5674MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5674MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5674MI Supplier MilliporeSigma Supplier No. MAB5674
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Plectin using this Anti-Plectin Antibody, Clone 10F6 validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plectin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 10F6
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3
Gene Symbols ACTC1, ACTG1, C14orf49, PLEC1
Host Species Mouse
Immunogen Gel-purified C5 Cell plectin
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.