Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pleiotrophin Mouse anti-Human, Clone: 3B10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC42
Description
This Anti-Pleiotrophin Antibody, clone 3B10 is validated for use in immunohistochemistry, western blotting for the detection of Pleiotrophin.
Specifications
Pleiotrophin | |
Monoclonal | |
Unconjugated | |
XP_416358 | |
Mouse | |
Protein G Purified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
3B10 | |
Purified mouse monoclonal IgG1κ in buffer containing PBS without 0.05% sodium azide. | |
OSF-1, PTN, NEGF1, HBNF-1, HBBM, HBGF8, HBGF-8, HBNF1, HB-GAM, HARP, HBNF1 | |
Recombinant protein corresponding to chicken Pleiotrophin. | |
50 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction