Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plexin B1 Mouse anti-Mouse, Rat, Human, Clone: 19A7.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB10402
Description
Anti-Plexin B1 Antibody, clone 19A7.1 detects level of Plexin B1 and has been published and validated for use in western blot, immunohistochemistry.
Specifications
Plexin B1 | |
Monoclonal | |
Unconjugated | |
NP_001123554 | |
Mouse | |
Ion Exchange | |
Primary | |
2-8°C for one year | |
IgM κ |
Immunohistochemistry, Western Blot | |
19A7.1 | |
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. | |
SEP, plexin 5, PLXN5, PLXNB1 | |
GST-tagged recombinant protein corresponding to human Plexin B1. | |
100 μg | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction