Learn More
PLIN, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PLIN peptide using ARM Technology.
Supplier: Abnova Corporation H00005346K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene coats lipid storage droplets in adipocytes, thereby protecting them until they can be broken down by hormone-sensitive lipase. The encoded protein is the major cAMP-dependent protein kinase substrate in adipocytes and, when unphosphorylated, may play a role in the inhibition of lipolysis. Alternatively spliced transcript variants varying in the 5' UTR, but encoding the same protein, have been found for this gene. [provided by RefSeq]
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PLIN | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PLIN | |
PLIN | |
A synthetic peptide of human PLIN | |
RUO | |
5346 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
perilipin | |
PLIN | |
PERI | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.