Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLK1 Mouse anti-Human, Mouse, Rat, Clone: 35-206, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05844
Description
Anti-PLK1 Antibody, clone 35-206 detects level of PLK1 and has been published and validated for use in western blot, immunoprecipitation, immunocytochemistry, immunohistochemistry.
Specifications
| PLK1 | |
| Monoclonal | |
| Unconjugated | |
| NP_005021 | |
| Mouse | |
| Protein A purfied | |
| Primary | |
| Stable for 1 year at from date of receipt. | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 35-206 | |
| Protein A purified mouse IgG2b in buffer containing 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide and 30% glycerol. Store at −20°C. | |
| PLK1, PLK, PLK-1, STPK13 | |
| 6His-tagged full length fusion protein corresponding to human polo-like kinase 1 (PLK1). Also called serine-threonine protein kinase PLK. | |
| 200 μg | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction