Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLXNA2, Mouse, Clone: 2G5, Abnova™

Catalog No. 89123266 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-266 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-266 Supplier Abnova Corporation Supplier No. H00005362M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant PLXNA2.

This gene encodes a member of the plexin-A family of semaphorin co-receptors. Semaphorins are a large family of secreted or membrane-bound proteins that mediate repulsive effects on axon pathfinding during nervous system development. A subset of semaphorins are recognized by plexin-A/neuropilin transmembrane receptor complexes, triggering a cellular signal transduction cascade that leads to axon repulsion. This plexin-A family member is thought to transduce signals from semaphorin-3A and -3C. [provided by RefSeq

Sequence: MHSQVFSAFPYSLPLRFWVNVIKNPQFVFDIHKGSITDACLSVVAQTFMDSCSTSEHRLDKDSPSNKLLYAKDIPSYKSWVERYYADIAKLPAISDQDMNAYLAEQSRLHAVEFNMLSALNEIYSYVSKYSEELIGALEQDEQARRQRLAYKVEQLINAMSIES

Specifications

Antigen PLXNA2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant PLXNA2.
Formulation PBS with no preservative; pH 7.4
Gene PLXNA2
Gene Accession No. BC032125
Gene Alias FLJ11751/FLJ30634/KIAA0463/OCT/PLXN2
Gene Symbols PLXNA2
Host Species Mouse
Immunogen PLXNA2 (AAH32125, 1 a.a. ∼ 164 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.