Learn More
PLZF Mouse, Alexa Fluor 647, Clone: R17-809, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563490
Description
The R17-809 monoclonal antibody recognizes promyelocytic leukemia zinc finger protein (PLZF). PLZF is also known as zinc finger and BTB domain containing 16 (ZBTB16) and Zinc finger protein 145. PLZF is a member of the BTB/POZ-ZF family of transcription factors that includes Th-POK (Zbtb7b) which is involved in CD4+ T cell fate determination. PLZF serves multiple functions. In the immune system, PLZF is involved in the developmental regulation of innate immune lymphocytes including NKT cells, a subset of γδ T cells, and mucosal associated invariant T (MAIT) cells. The R17-809 hybridoma was generated from a mouse immunized with recombinant mouse PLZF protein. The R17-809 antibody crossreacts with both mouse and human PLZF.

Specifications
PLZF | |
Monoclonal | |
0.2mg/mL | |
Zbtb16; zinc finger and BTB domain containing 16; zinc finger protein 145 | |
Mouse | |
Affinity Purified | |
RUO | |
Human, Mouse | |
IgG1 κ |
Flow Cytometry | |
R17-809 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse PLZF Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.