Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polyamine modulated factor 1 binding protein 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008281 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-281 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-281 Supplier Abnova Corporation Supplier No. H00083449B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PMFBP1 protein.

Sequence: MKDEAGERDREVSSLNSKLLSLQLDIKNLHDVCKRQRKTLQDNQLCMEEAMNSSHDKKQAQALAFEESEVEFGSSKQCHLRQLQQLKKKLLVLQQELEFHTEELQTSYYSLRQYQSILEKQTSDLVLLHHHCKLKEDEVILYEEEMGNHNENTGEKLHLAQEQLALAGDKIASLERSLNLYRDKYQSSLSNIELLECQVKMLQGELGGIMGQEPENKGDHSKVRIYTSPCMIQEHQETQKRLSEVWQKVSQQDDLIQELRNKLACSNALVLEREKALIKLQADFASCTATHRYPPSSSEECEDIKKILKHLQEQKDSQCLHVEEYQNLVKDLRVELEAVSEQKRNIMKDMMKLELDLHGLREETSAHIERKDKDITILQCRLQELQLEFTETQKLTLKKDKFLQEKDEMLQELEKKLTQVQNSLLKKEKELEKQQCMATELEMTVKEAKQDKSKEAECKALQAEVQKLKNSLEEAKQQERLAGEAPAAQQAAQCKEEAALAGCHLEDTQRKLQKGLLLDKQKADTIQELQRELQMLQKESSMAEKEQTSNRKRVEELSLELSEALRKLENSDKEKRQLQKTVAEQDMKMNDMLDRIKHQHREQGSIKCKLEEDLQEATKLLEDKREQLKKSKEHEKLMEGELEALRQEFKKKDKTLKENSRKLEEENENLRAELQCCSTQLESSLNKYNTSQQVIQDLNKEIALQKESLMSLQAQLDKALQKEKHYLQTTITKEAYDALSRKSAACQDDLTQALEKLNHVTSETKSLQQSLTQTQEKKAQLEEEIIAYEERMKKLNTELRKLRGFHQESELEVHAFDKKLEEMSCQVLQWQKQHQNDLKMLAAKEEQLREFQEEMAALKENLLEDDKEPCCLPQWSVPKDTCRLYRGNDQIMTNLEQWAKQQKVANEKLGNQLREQVKYIAKLSGEKDHLHSVMVHLQQENKKLKKEIEEKKMKAENTRLCTKALGPSRTESTQREKVCGTLGWKGLPQDMGQRMDLTKYIGMPHCPGTSAIGQKNKCDFFL

Specifications

Antigen polyamine modulated factor 1 binding protein 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PMFBP1 protein.
Formulation No additive
Gene PMFBP1
Gene Accession No. BC028423.2
Gene Alias DKFZp434G131/FLJ40146
Gene Symbols PMFBP1
Host Species Mouse
Immunogen PMFBP1 (AAH28423.1, 1 a.a. ∼ 1022 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 83449
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.