Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PML Mouse anti-Mouse, Clone: 36.1-104, MilliporeSigma™

Catalog No. 05718MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-718-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-718-MI Supplier MilliporeSigma Supplier No. 05718
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect PML also known as Promyelocytic Leukemia Protein with Anti-PML Antibody, clone 36.1-104 (Mouse Monoclonal Antibody), that has been demonstrated to work in immunocytochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PML
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 36.1-104
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative.
Gene Accession No. NM_033238
Gene Symbols PML, PP8675, RNF71, TRIM19, MYL
Host Species Mouse
Immunogen 6His fusion protein corresponding to mouse PML (promyelocytic leukemia protein)
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.