Learn More
PMS2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human PMS2 peptide using ARM Technology.
Supplier: Abnova Corporation H00005395K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
PMS2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
PMS2 | |
PMS2 | |
A synthetic peptide of human PMS2 | |
RUO | |
5395 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
PMS2 postmeiotic segregation increased 2 (S. cerevisiae) | |
PMS2 | |
HNPCC4, PMS2CL, PMSL2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.