Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMS2 postmeiotic segregation increased 2 (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005975 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-975 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-975 Supplier Abnova Corporation Supplier No. H00005395B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human PMS2 protein.

This gene is one of the PMS2 gene family members found in clusters on chromosome 7. The product of this gene is involved in DNA mismatch repair. It forms a heterodimer with MLH1 and this complex interacts with other complexes bound to mismatched bases. Mutations in this gene are associated with hereditary nonpolyposis colorectal cancer, Turcot syndrome, and are a cause of supratentorial primitive neuroectodermal tumors. Alternatively spliced transcript variants have been observed for this gene. [provided by RefSeq

Sequence: MERAESSSTEPAKAIKPIDRKSVHQICSGQVVLSLSTAVKELVENSLDAGATNIDLKLKDYGVDLIEVSDNGCGVEEENFEGLTLKHHTSKIQEFADLTQVETFGFRGEALSSLCALSDVTISTCHASAKVGTRLMFDHNGKIIQKTPYPRPRGTTVSVQQLFSTLPVRHKEFQRNIKKEYAKMVQVLHAYCIISAGIRVSCTNQLGQGKRQPVVCTGGSPSIKENIGSVFGQKQLQSLIPFVQLPPSDSVCEEYGLSCSDALHNLFYISGFISQCTHGVGRSSTDRQFFFINRRPCDPAKVCRLVNEVYHMYNRHQYPFVVLNISVDSECVDINVTPDKRQILLQEEKLLLAVLKTSLIGMFDSDVNKLNVSQQPLLDVEGNLIKMHAADLEKPMVEKQDQSPSLRTGEEKKDVSISRLREAFSLRHTTENKPHSPKTPEPRRSPLGQKRGMLSSSTSGAISDKGVLRPQKEAVSSSHGPSDPTDRAEVEKDSGHGSTSVDSEGFSIPDTGSHCSSEYAASSPGDRGSQEHVDSQEKAPETDDSFSDVDCHSNQEDTGCKFRVLPQPTNLATPNTKRFKKEEILSSSDICQKLVNTQDMSASQVDVAVKINKKVVPLDFSMSSLAKRIKQLHHEAQQSEGEQNYRKFRAKICPGENQAAEDELRKEISKTMFAEMEIIGQFNLGFIITKLNEDIFIVDQHATDEKYNFEMLQQHTVLQGQRLIAPQTLNLTAVNEAVLIENLEIFRKNGFDFVIDENAPVTERAKLISLPTSKNWTFGPQDVDELIFMLSDSPGVMCRPSRVKQMFASRACRKSVMIGTALNTSEMKKLITHMGEMDHPWNCPHGRPTMRHIANLGVISQN

Specifications

Antigen PMS2 postmeiotic segregation increased 2 (S. cerevisiae)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human PMS2 protein.
Formulation No additive
Gene PMS2
Gene Accession No. NM_000535
Gene Alias HNPCC4/PMS2CL/PMSL2
Gene Symbols PMS2
Host Species Mouse
Immunogen PMS2 (NP_000526, 1 a.a. ∼ 862 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5395
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.