Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

paraneoplastic antigen MA1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-651 Supplier Abnova Corporation Supplier No. H00009240D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human PNMA1 protein.

Sequence: MAMTLLEDWCRGMDVNSQRALLVWGIPVNCDEAEIEETLQAAMPQVSYRMLGRMFWREENAKAALLELTGAVDYAAIPREMPGKGGVWKVLFKPPTSDAEFLERLHLFLAREGWTVQDVARVLGFQNPTPTPGPEMPAEMLNYILDNVIQPLVESIWYKRLTLFSGRDIPGPGEETFDPWLEHTNEVLEEWQVSDVEKRRRLMESLRGPAADVIRILKSNNPAITTAECLKALEQVFGSVESSRDAQIKFLNTYQNPGEKLSAYVIRLEPLLQKVVEKGAIDKDNVNQARLEQVIAGANHSGAIRRQLWLTGAGEGPAPNLFQLLVQIREEEAKEEEEEAEATLLQLGLEGHF

Specifications

Antigen paraneoplastic antigen MA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human PNMA1 protein.
Formulation PBS with no preservative; pH 7.4
Gene PNMA1
Gene Accession No. NM_006029.4
Gene Alias MA1
Gene Symbols PNMA1
Host Species Rabbit
Immunogen PNMA1 (NP_006020.4, 1 a.a. ∼ 353 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 9240
Target Species Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.