Learn More
phenylethanolamine N-methyltransferase (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant PNMT.
Supplier: Abnova Corporation H00005409A01
Description
The product of this gene catalyzes the last step of the catecholamine biosynthesis pathway, which methylates norepinephrine to form epinephrine (adrenaline). The enzyme also has beta-carboline 2N-methyltransferase activity. This gene is thought to play a key step in regulating epinephrine production. [provided by RefSeq
Sequence: MSGADRSPNAGAAPDSAPGQAAVASAYQRFEPRAYLRNNYAPPRGDLCNPNGVGPWKLRCLAQTFATGEVSGRTLIDIGSGPTVYQLLSACSHFEDITMTDFLEVNRQELSpecifications
phenylethanolamine N-methyltransferase | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant PNMT. | |
PNMT | |
MGC34570/PENT/PNMTase | |
Mouse | |
50 μL | |
Yes | |
5409 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_002686 | |
PNMT | |
PNMT (NP_002677, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.