Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
patatin-like phospholipase domain containing 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human PNPLA2 protein.
Supplier: Abnova Corporation H00057104D01P
Description
Sequence: MFPREKTWNISFAGCGFLGVYYVGVASCLREHAPFLVANATHIYGASAGALTATALVTGVCLGEAGAKFIEVSKEARKRFLGPLHPSFNLVKIIRSFLLKVLPADSHEHASGRLGISLTRVSDGENVIISHFNSKDELIQANVCSGFIPVYCGLIPPSLQGVRYVDGGISDNLPLYELKNTITVSPFSGESDICPQDSSTNIHELRVTNTSIQFNLRNLYRLSKALFPPEPLVLREMCKQGYRDGLRFLQRNGLLNRPNPLLALPPARPHGPEDKDQAVESAQAEDYSQLPGEDHILEHLPARLNEALLEACVEPTDLLTTLSNMLPVRLATAMMVPYTLPLESALSFTIRLLEWLPDVPEDIRWMKEQTGSICQYLVMRAKRKLGRHLPSRLPEQVELRRVQSLPSVPLSCAAYREALPGWMRNNLSLGDALAKWEECQRQLLLGLFCTNVAFPPEALRMRAPADPAPAPADPASPQHQLAGPAPLLSTPAPEARPVIGALGLSpecifications
patatin-like phospholipase domain containing 2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human PNPLA2 protein. | |
PNPLA2 | |
1110001C14Rik/ATGL/DESNUTRIN/DKFZp667M109/FP17548/PEDF-R/TTS-2.2/TTS2 | |
Rabbit | |
Affinity Purified | |
RUO | |
57104 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_020376.2 | |
PNPLA2 | |
PNPLA2 (NP_065109.1, 1 a.a. ∼ 504 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction