Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podocalyxin-like protein I Mouse anti-Human, Clone: 85, MilliporeSigma™

Catalog No. MAB4433MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4433MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4433MI Supplier MilliporeSigma Supplier No. MAB4433

Mouse Monoclonal Antibody

Detect Podocalyxin-like protein I using this Anti-Podocalyxin-like protein I Antibody, clone 85 validated for use in immunocytochemistry, FC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podocalyxin-like protein I
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 85
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001018121
Gene Symbols PCLP, Gp200, PC, PCLP1
Host Species Mouse
Immunogen Human embryonic stem cells.
Purification Method Ion exchange purification column
Quantity 200 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.