Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Podoplanin Rat anti-Human, Clone: NZ1.2, MilliporeSigma™

Catalog No. MABC117MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC117MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC117MI Supplier MilliporeSigma Supplier No. MABC117
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-Podoplanin Antibody, clone NZ1.2 is an antibody against Podoplanin for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Podoplanin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone NZ1.2
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001006625
Gene Symbols PDPN, GP36, GP36, PSEC0003, PSEC0025
Host Species Rat
Immunogen Protein sequence corresponding to the PLAG domain of human Podoplanin.
Purification Method Protein G Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.