Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polymerase (DNA-directed), delta 4, Mouse, Clone: 2C11, Abnova™

Catalog No. 89001330 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-330 Supplier Abnova Corporation Supplier No. H00057804M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant POLD4.

The DNA polymerase delta complex is involved in DNA replication and repair, and it consists of the proliferating cell nuclear antigen (PCNA; MIM 176740), the multisubunit replication factor C (see MIM 102579), and the 4 subunit polymerase complex: POLD1 (MIM 174761), POLD2 (MIM 600815), POLD3 (MIM 611415), and POLD4 (Liu and Warbrick, 2006 [PubMed 16934752]).[supplied by OMIM

Sequence: MGRKRLITDSYPVVKRREGPAGHSKGELAPELGL

Specifications

Antigen polymerase (DNA-directed), delta 4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant POLD4.
Formulation PBS with no preservative; pH 7.4
Gene POLD4
Gene Accession No. BC001334
Gene Alias POLDS/p12
Gene Symbols POLD4
Host Species Mouse
Immunogen POLD4 (AAH01334, 1 a.a. ∼ 34 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57804
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.