Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polymerase (RNA) I polypeptide B, 128kDa, Mouse, Clone: 4H6, Abnova™

Catalog No. 89001371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-371 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-371 Supplier Abnova Corporation Supplier No. H00084172M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant POLR1B.

Eukaryotic RNA polymerase I (pol I) is responsible for the transcription of ribosomal RNA (rRNA) genes and production of rRNA, the primary component of ribosomes. Pol I is a multisubunit enzyme composed of 6 to 14 polypeptides, depending on the species. Most of the mass of the pol I complex derives from the 2 largest subunits, Rpa1 and Rpa2 in yeast. POLR1B is homologous to Rpa2 (Seither and Grummt, 1996 [PubMed 8921381]).[supplied by OMIM

Sequence: GEMLKAAGYNFYGTERLYSGISGLELEADIFIGVVYYQRLRHMVSDKFQVRTTGARDRVTNQPIGGRNVQGGIRFGEMERDALLAHGTSFLLHDRLFNCSDRSVAHVCVK

Specifications

Antigen polymerase (RNA) I polypeptide B, 128kDa
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant POLR1B.
Formulation PBS with no preservative; pH 7.4
Gene POLR1B
Gene Accession No. NM_019014
Gene Alias FLJ10816/FLJ21921/MGC131780/RPA135/RPA2/Rpo1-2
Gene Symbols POLR1B
Host Species Mouse
Immunogen POLR1B (NP_061887, 963 a.a. ∼ 1072 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84172
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.