Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

polymerase (RNA) I polypeptide C, 30kDa, Mouse, Clone: M1, Abnova™

Catalog No. 89001141 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-141 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-141 Supplier Abnova Corporation Supplier No. H00009533M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant POLR1C.

Sequence: MAASQAVEEMRSRVVLGEFGVRNVHTTDFPGNYSGYDDAWDQDRFEKNFRVDVVHMDENSLEFDMVGIDAAIANAFRRILLAEVPTMAVEKVLVYNNTSIVQDEILAHRLGLIPIHADPRLFEYRNQGDEEGTEIDTLQFRLQVRCTRNPHAAKDSSDPNELYVNHKVYTRHMTWIPLGNQADLFPEGTIRPVHDDILIAQLRPGQEIDLLMHCVKGIGKDHAKFSPVATASYRLLPDITLLEPVEGEAAEELSRCFSPGVIEVQEVQGKKVARVANPRLDTFSREIFRNEKLKKVVRLARVRDHYIFSVESTGVLPPDVLVSEAIKVLMGKCRRFLDELDAVQMD

Specifications

Antigen polymerase (RNA) I polypeptide C, 30kDa
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone M1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant POLR1C.
Formulation PBS with no preservative; pH 7.4
Gene POLR1C
Gene Accession No. BC008863
Gene Alias RPA39/RPA40/RPA5/RPAC1
Gene Symbols POLR1C
Host Species Mouse
Immunogen POLR1C (AAH08863, 1 a.a. ∼ 346 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9533
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.